Learn More
Medchemexpress LLC SUN 1334H | 607736-84-5 | 489.38 | 1 MG
SDP
Catalog No. 501934410
Encompass_Preferred
SUN 1334H is a potent, orally active, and highly selective H1 receptor antagonist with a Ki of 9.7 nM. It causes potent inhibition of histamine-induced contractions and has been shown to inhibit histamine-induced bronchospasm and skin wheal. It demonstrates no significant central nervous system side effects.
  • Potent, orally active, and highly selective H1 receptor antagonist
  • Inhibits histamine-induced contractions of isolated guinea-pig ileum
  • Does not modulate hERG K+-currents at high concentrations
  • Inhibits histamine-induced bronchospasm and skin wheal
  • Reduces passive and active cutaneous anaphylactic reactions
  • Devoid of significant central nervous system side effects
Catalog No. 50-193-4410 Supplier Medchemexpress LLC Supplier No. HYU000841MG
SUN 1334H is a potent, orally active, and highly selective H1 receptor antagonist with a Ki of 9.7 nM. It causes potent inhibition of histamine-induced contractions and has been shown to inhibit histamine-induced bronchospasm and skin wheal. It demonstrates no significant central nervous system side effects.
  • Potent, orally active, and highly selective H1 receptor antagonist
  • Inhibits histamine-induced contractions of isolated guinea-pig ileum
  • Does not modulate hERG K+-currents at high concentrations
  • Inhibits histamine-induced bronchospasm and skin wheal
  • Reduces passive and active cutaneous anaphylactic reactions
  • Devoid of significant central nervous system side effects