Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Sun-Earth-Moon Flip Chart with Online Multimedia Lesson
SCI_ED

Catalog No. S81125 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S81125 6 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81125 Supplier NewPath Learning™ Supplier No. 346850
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Combine illuminating graphics and write-on/wipe-off exercises with multimedia lessons to master key earth science topics.

  • Provides graphical instruction and write-on/wipe-off activities with one-year online access to a corresponding Multimedia Lesson featuring in-depth, interactive coverage of the concept
  • The Multimedia Lesson can be accessed by a teacher and ALL of your students anywhere, anytime, on any device – including Chromebooks, tablets, computers and mobile devices.
  • Flip Chart Set features 10 Laminated, Double-Sided Charts
  • Side 1 features a graphic overview of the topic while Side 2 provides corresponding write-on/wipe-off activities for review and reinforcement
  • Includes a Teacher Guide with copymaster activities and review questions
  • Spiral-bound and mounted on a sturdy easel for years of use, the Flip Chart sets are ideal for small group instruction, learning centers, independent use or classroom reference
  • Also included is one year access to a corresponding online multimedia lesson for a teacher and all their students
  • Each lesson features Animated, Narrated Tutorials; Interactive Exercises & Activities; Virtual Lab Investigations; Illustrated, Narrated Glossary; Visual Resource Library; and an Inquiry-Based Student Activity Guide
  • Developed under sponsorship of the National Institutes of Health (NIH), the lessons are perfect for individual student instruction on any computer as well as classroom presentation on any Interactive Whiteboard
  • Covers: How the Earth Moves; Earth’s Hemispheres; Seasons on Earth; Gravity & Motion; Earth’s Moon; Phases of the Moon; Eclipses; Tides; and Missions to the Moon

Specifications

Grade Level Intermediate - Middle School, High School
Grade 6 to 10
Description NGSS Flip Chart with Online Lesson
Category Earth Science
Format Download
For Use With (Application) Classroom Instruction
Includes Flip Chart, Online Lesson
License Site
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.