Learn More
Thermo Scientific™ SUN-SRi™ 8mm Vial Crimp Seals
Catalog No. 14823230 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Natural
Silver
Septum:
None
PTFE/Silicone/PTFE
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum
14-823-230 Silver PTFE/Silicone/PTFE
14-823-424 Silver None
14-823-425 Natural None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-823-230 Supplier Thermo Scientific™ Supplier No. 201 876
Get an Extra Case of Product with Purchase
View all promotions
Add to Cart
Add to Cart
Thermo Scientific caps and vials are available in a wide variety of sizes and configurations to fit the needs of many applications.
TRUSTED_SUSTAINABILITY
Specifications
Color Silver
Material Aluminum
Pre-slit No
Septum PTFE/Silicone/PTFE
Quantity 1000 Cs.
Closure Size 8 mm
Product Type Crimp Cap