Learn More
Thermo Scientific™ SUN-SRi™ Aluminum Seals for 20mm Serum Bottles 1000 per cs
Shop All Thermo Scientific ProductsSpecifications
Specifications
| Material | Aluminum |
| Style | Standard Opening |
| Quantity | 1000 Cs. |
| Closure Size | 20 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.