Learn More
MSC Sun Tec Corp Hardness Calibration Test Block, Rockwell C, C25 Hardness, Low Range
Catalog No. 5000513285
Encompass
This hardness calibration test block is designed for precise measurements on the Rockwell C scale. Featuring a C25 hardness, it is ideal for calibrating instruments within the low hardness range, ensuring accuracy and reliability in testing applications.
  • Block type: Test
  • Scale type: Rockwell C
  • Maximum hardness: 25 HRC
  • Hardness range description: Low
  • Made in the USA
  • TAA compliant
  • Section 889 compliant
Catalog No. 50-005-13285 Supplier MSC Supplier No. 00444711
This hardness calibration test block is designed for precise measurements on the Rockwell C scale. Featuring a C25 hardness, it is ideal for calibrating instruments within the low hardness range, ensuring accuracy and reliability in testing applications.
  • Block type: Test
  • Scale type: Rockwell C
  • Maximum hardness: 25 HRC
  • Hardness range description: Low
  • Made in the USA
  • TAA compliant
  • Section 889 compliant