Learn More
MSC Sun Tec Corp Hardness Calibration Test Block, Rockwell C, Steel, C45
Catalog No. 5000529666
Encompass
This hardness calibration test block is designed for precise measurements on the Rockwell C scale. Manufactured from durable steel, it features a C45 hardness, suitable for a wide range of industrial applications.
  • Calibrated to Rockwell C scale.
  • Made from robust steel.
  • Features C45 hardness for mid-range testing.
  • Includes NIST traceable calibration certificates.
  • Manufactured in the United States.
  • TAA compliant.
  • Section 889 compliant.
Catalog No. 50-005-29666 Supplier MSC Supplier No. 00444794
This hardness calibration test block is designed for precise measurements on the Rockwell C scale. Manufactured from durable steel, it features a C45 hardness, suitable for a wide range of industrial applications.
  • Calibrated to Rockwell C scale.
  • Made from robust steel.
  • Features C45 hardness for mid-range testing.
  • Includes NIST traceable calibration certificates.
  • Manufactured in the United States.
  • TAA compliant.
  • Section 889 compliant.