Learn More
MSC Sunex Tools Portable Hydraulic Jack, 4 Ton Load Capacity, Steel
Catalog No. 5000513599
Encompass
This portable hydraulic jack kit is designed for a variety of applications, including pushing, spreading, bending, collision repair, and clamping. It features a 4-ton load capacity and is constructed from durable steel.
  • Portable hydraulic jack type
  • 4 ton load capacity
  • Minimum height of 10.50 in
  • Power raise of 4.72 in
  • Suitable for pushing, spreading, bending, collision repair work, and clamping
  • Constructed from steel
  • Comes as a 13 piece kit
Catalog No. 50-005-13599 Supplier MSC Supplier No. 00445064
This portable hydraulic jack kit is designed for a variety of applications, including pushing, spreading, bending, collision repair, and clamping. It features a 4-ton load capacity and is constructed from durable steel.
  • Portable hydraulic jack type
  • 4 ton load capacity
  • Minimum height of 10.50 in
  • Power raise of 4.72 in
  • Suitable for pushing, spreading, bending, collision repair work, and clamping
  • Constructed from steel
  • Comes as a 13 piece kit