Learn More
Waters Corp SunFire C18 Column, 10 μm, 4.6 mm ID, 150 mm Length
Catalog No. 50465955 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds, and is ideally suited for purification and impurity profile assays.
  • C18 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range from 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 10 μm particle size
  • 100 Å pore size
  • Column format
  • 340 surface area
  • HPLC system compatible
  • L1 USP classification
  • 4.6 mm inner diameter
  • 150 mm length
  • 16% carbon load
  • 1 unit per package
Catalog No. 50-465-955 Supplier Waters Corp Supplier No. 186003390
SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds, and is ideally suited for purification and impurity profile assays.
  • C18 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range from 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 10 μm particle size
  • 100 Å pore size
  • Column format
  • 340 surface area
  • HPLC system compatible
  • L1 USP classification
  • 4.6 mm inner diameter
  • 150 mm length
  • 16% carbon load
  • 1 unit per package