Learn More
Waters Corp SunFire C18 Column, 100Å, 10 μm, 4.6 mm × 250 mm
Supplier:  Waters Corp 186003391
Encompass_Preferred
SunFire C18 columns are used for general-purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds, making it ideal for purification and impurity profile assays. These columns aid in developing general-purpose methods at low pH and combine particle and bonding expertise for industry-leading chromatographic performance.
  • High resolution
  • Fast speed
  • Low back pressure
  • Excellent sensitivity
  • High peak capacity
  • Ultra-low bleed
  • Can be used in a variety of mass spectrometry applications
  • Packing materials developed in a cGMP, ISO 9002-certified facility using ultra-pure reagent
Catalog No. 50-781-729