Learn More
Waters Corp Waters SunFire C18 Column, 100Å, 2.5 μm, 2.1 mm × 30 mm
Supplier:  Waters Corp 186003399
Encompass_Preferred
SunFire C18 columns are designed for general purpose method development in low pH environments. They feature a highly retentive C18 ligand, which is particularly effective for basic compounds, making them suitable for purification and impurity profile assays.
  • Used for general purpose method development at low pH.
  • Highly retentive C18 ligand, especially for basic compounds.
  • Ideally suited for purification assays.
  • Ideally suited for impurity profile assays.
Catalog No. 50-781-736