Learn More
Waters Corp Waters Corporation SunFire Column, C18, 100Å, 2.5 μm, 3 mm × 75 mm
Catalog No. 50466169 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds, making them ideally suited for purification and impurity profile assays. These advanced bonded C18 silica HPLC columns offer industry-leading chromatographic performance, combining expertise in particle and bonding for high efficiency, low pH stability, and better peak morphologies for charged analyte species.
  • High chromatographic efficiency
  • Low pH stability
  • Better peak morphologies for charged analyte species
  • High resolution
  • Quickness
  • Low back pressure
  • Exceptional sensitivity
  • Big peak capacity
  • Extremely little bleed, resulting in sharp peaks
  • Suitable for a variety of mass spectrometry applications
  • Enhanced column lifetime due to lower pH stability compared to normal silica-based HPLC columns
Catalog No. 50-466-169 Supplier Waters Corp Supplier No. 186005636
SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds, making them ideally suited for purification and impurity profile assays. These advanced bonded C18 silica HPLC columns offer industry-leading chromatographic performance, combining expertise in particle and bonding for high efficiency, low pH stability, and better peak morphologies for charged analyte species.
  • High chromatographic efficiency
  • Low pH stability
  • Better peak morphologies for charged analyte species
  • High resolution
  • Quickness
  • Low back pressure
  • Exceptional sensitivity
  • Big peak capacity
  • Extremely little bleed, resulting in sharp peaks
  • Suitable for a variety of mass spectrometry applications
  • Enhanced column lifetime due to lower pH stability compared to normal silica-based HPLC columns