Learn More
Waters Corp Waters Column, C18, 2.5 μm, 3 mm x 50 mm, 100 Å
Catalog No. 50781746 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These C18 columns provide high-resolution, fast, and low-back pressure performance for general purpose method development at low pH. Suitable for various mass spectrometry applications, they offer excellent low pH stability, extending column lifetime.
  • High-resolution, fast, and low-back pressure performance.
  • Exceptional sensitivity and large peak capacity.
  • Minimal bleed, resulting in strong peaks.
  • Highly retentive C18 ligand, effective for basic compounds.
  • Ideally suited for purification and impurity profile assays.
  • Manufactured with ultra-pure reagents in a cGMP, ISO 9001 certified facility.
  • Batches are chromatographically tested for outstanding, repeatable performance.
  • Superior peak shape with symmetrical peaks for better resolution and quantification.
Catalog No. 50-781-746 Supplier Waters Corp Supplier No. 186003409
Add to Cart
Add to Cart
These C18 columns provide high-resolution, fast, and low-back pressure performance for general purpose method development at low pH. Suitable for various mass spectrometry applications, they offer excellent low pH stability, extending column lifetime.
  • High-resolution, fast, and low-back pressure performance.
  • Exceptional sensitivity and large peak capacity.
  • Minimal bleed, resulting in strong peaks.
  • Highly retentive C18 ligand, effective for basic compounds.
  • Ideally suited for purification and impurity profile assays.
  • Manufactured with ultra-pure reagents in a cGMP, ISO 9001 certified facility.
  • Batches are chromatographically tested for outstanding, repeatable performance.
  • Superior peak shape with symmetrical peaks for better resolution and quantification.