Learn More
Waters Corp Waters HPLC Column, C18, 2.5 μm, 3 mm ID, 30 mm Length
Catalog No. 50781744 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C18 columns are designed for general purpose method development at low pH. The C18 ligand provides high retention, particularly for basic compounds, making it suitable for purification and impurity profile assays.
  • Impeccable peak shape and remarkable efficiency due to its 2.5μm particle size.
  • Strong retention of basic compounds enabled by the C18 bonded phase, allowing separation of a wide range of analytes with precision and accuracy.
  • End-capped to minimize column bleed and optimize peak shape, leading to cleaner chromatograms and improved data integrity.
  • Compatibility with various solvents such as water, acetonitrile, methanol, and tetrahydrofuran.
  • Operating temperatures up to 45°C and ability to withstand pressures up to 6000 psi, making it suitable for demanding conditions and high-temperature applications.
  • Recommended pH range between 2 and 8.
Catalog No. 50-781-744 Supplier Waters Corp Supplier No. 186003407
SunFire C18 columns are designed for general purpose method development at low pH. The C18 ligand provides high retention, particularly for basic compounds, making it suitable for purification and impurity profile assays.
  • Impeccable peak shape and remarkable efficiency due to its 2.5μm particle size.
  • Strong retention of basic compounds enabled by the C18 bonded phase, allowing separation of a wide range of analytes with precision and accuracy.
  • End-capped to minimize column bleed and optimize peak shape, leading to cleaner chromatograms and improved data integrity.
  • Compatibility with various solvents such as water, acetonitrile, methanol, and tetrahydrofuran.
  • Operating temperatures up to 45°C and ability to withstand pressures up to 6000 psi, making it suitable for demanding conditions and high-temperature applications.
  • Recommended pH range between 2 and 8.