Learn More
Waters Corp Waters SunFire C18 Column, 100Å, 3.5 μm, 2.1 × 100 mm
Catalog No. 50781174 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C18 columns are ideal for general purpose method development at low pH. Highly retentive, they excel in purification and impurity profile assays, offering industry-leading chromatographic performance through advanced particle and bonding.
  • Highly retentive C18 ligand for basic compounds.
  • Suitable for purification and impurity profile assays.
  • Delivers industry-leading chromatographic performance.
  • Offers high efficiency, low pH stability, and improved peak morphologies.
  • Provides high resolution, speed, and low backpressure.
  • Ensures outstanding sensitivity, large capacity, and ultra-low bleed for sharp peaks.
  • Wide applicability in mass spectrometry.
  • Increased column’s lifetime from low pH stability.
  • Innovative packaging and advanced silica synthesis for remarkable sensitivity.
Catalog No. 50-781-174 Supplier Waters Corp Supplier No. 186002534
SunFire C18 columns are ideal for general purpose method development at low pH. Highly retentive, they excel in purification and impurity profile assays, offering industry-leading chromatographic performance through advanced particle and bonding.
  • Highly retentive C18 ligand for basic compounds.
  • Suitable for purification and impurity profile assays.
  • Delivers industry-leading chromatographic performance.
  • Offers high efficiency, low pH stability, and improved peak morphologies.
  • Provides high resolution, speed, and low backpressure.
  • Ensures outstanding sensitivity, large capacity, and ultra-low bleed for sharp peaks.
  • Wide applicability in mass spectrometry.
  • Increased column’s lifetime from low pH stability.
  • Innovative packaging and advanced silica synthesis for remarkable sensitivity.