Learn More
Waters Corp Waters Column, C18, 3.5 μm, 100 Å, 3 mm x 50 mm
Supplier:  Waters Corp 186002542
Encompass_Preferred
SunFire C18 columns are designed for general-purpose method development at low pH. The C18 ligand provides high retention, particularly for basic compounds, making it suitable for purification and impurity profile assays.
  • Achieves high resolution, speed, and low backpressure.
  • Provides outstanding sensitivity, large peak capacity, and ultra-low bleed for sharp peaks.
  • Compatible with various mass spectrometry applications.
  • Offers exceptional low pH stability, extending column lifetime.
  • Features proprietary silica synthesis, bonding, end-capping, and packing technologies.
  • Generates superior peak shape performance through advanced sorbent and bonding technology.
  • Ensures symmetrical peaks for improved resolution and quantitation of acidic, neutral, and basic compounds.
  • Operates effectively within a 2 – 8 pH range.
Catalog No. 50-781-182