Learn More
Waters Corp Waters Column, C18, 3.5 μm, 3 mm × 100 mm, 100 Å
Catalog No. 50781183 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
C18 columns provide high-resolution, fast, and low-back pressure performance with exceptional sensitivity, suitable for general purpose method development at low pH and mass spectrometry applications. They offer high retention for basic compounds and extended column lifetime due to enhanced low pH stability.
  • High-resolution, fast, low-back pressure.
  • Exceptional sensitivity.
  • Versatile for mass spectrometry.
  • General purpose method development at low pH.
  • High retention for basic compounds.
  • Extended column lifetime.
  • Proprietary technologies.
  • Effective across 2 to 8 pH range.
Catalog No. 50-781-183 Supplier Waters Corp Supplier No. 186002543
C18 columns provide high-resolution, fast, and low-back pressure performance with exceptional sensitivity, suitable for general purpose method development at low pH and mass spectrometry applications. They offer high retention for basic compounds and extended column lifetime due to enhanced low pH stability.
  • High-resolution, fast, low-back pressure.
  • Exceptional sensitivity.
  • Versatile for mass spectrometry.
  • General purpose method development at low pH.
  • High retention for basic compounds.
  • Extended column lifetime.
  • Proprietary technologies.
  • Effective across 2 to 8 pH range.