Learn More
Waters Corp Waters SunFire Column, C18, 100Å, 3.5 μm, 4.6 mm x 150 mm
Supplier:  Waters Corp 186002554
Encompass_Preferred
SunFire C18 columns are designed for general-purpose method development at low pH. The C18 ligand is highly retentive for basic compounds, making them suitable for purification and impurity profile assays. These columns offer excellent peak shape and high mass loading, making them a reliable choice for analytical and preparative needs.
  • General purpose method development at low pH.
  • Highly retentive C18 ligand for basic compounds.
  • Extended lifetime due to low pH stability.
  • High efficiency and greater sensitivity from proprietary silica technology.
  • Sharp peaks, excellent sensitivity, and ultra-low bleed.
  • Symmetrical peaks improve resolution and quantitation of compounds.
  • Chromatographically tested for reproducibility.
  • Reliable for analytical methods in pharmaceutical and biopharmaceutical products.
Catalog No. 50-781-194