Learn More
Waters Corp Waters Corporation SunFire C18 Column, 100Å 3.5 μm, 4.6 × 100 mm
Catalog No. 50781193 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Corporation SunFire C18 Column, 100Å 3.5 μm, 4.6 × 100 mm
Catalog No. 50-781-193 Supplier Waters Corp Supplier No. 186002553
Add to Cart
Add to Cart
Waters Corporation SunFire C18 Column, 100Å 3.5 μm, 4.6 × 100 mm