Learn More
Waters Corp Waters Corporation C18 Column, 100Å, 5 μm, 2.1 mm X 50 mm
Catalog No. 50781179 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C18 columns are engineered for general purpose method development at low pH. These columns feature a highly retentive C18 ligand, making them particularly suitable for basic compounds. They are ideally suited for applications such as purification and impurity profile assays.
  • General purpose method development at low pH
  • Highly retentive C18 ligand, especially for basic compounds
  • Ideal for purification and impurity profile assays
Catalog No. 50-781-179 Supplier Waters Corp Supplier No. 186002539
SunFire C18 columns are engineered for general purpose method development at low pH. These columns feature a highly retentive C18 ligand, making them particularly suitable for basic compounds. They are ideally suited for applications such as purification and impurity profile assays.
  • General purpose method development at low pH
  • Highly retentive C18 ligand, especially for basic compounds
  • Ideal for purification and impurity profile assays