Learn More
Waters Corp SunFire C18 Column, 100Å, 5 μm, 3 mm × 150 mm
Catalog No. 50781187 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C18 columns are ideal for general purpose method development at low pH, purification, and impurity assays. They offer high chromatographic performance, manufactured with stringent quality control for reliability and effectiveness.
  • High-resolution, quick separation, and low back-pressure.
  • Excellent sensitivity, large peak capacity, and minimal bleed for sharp peaks.
  • Adaptable for mass spectrometry applications; C18 ligand offers high retention for basic compounds.
  • Withstands maximum pressure of 6000 psi (415 Bar) with superior low pH stability.
  • Features proprietary silica synthesis, bonding, and end-capping procedures, improving peak shape and symmetry for resolution and quantitation of acidic, neutral, and basic compounds at 2 to 8 pH range.
Catalog No. 50-781-187 Supplier Waters Corp Supplier No. 186002547
Add to Cart
Add to Cart
SunFire C18 columns are ideal for general purpose method development at low pH, purification, and impurity assays. They offer high chromatographic performance, manufactured with stringent quality control for reliability and effectiveness.
  • High-resolution, quick separation, and low back-pressure.
  • Excellent sensitivity, large peak capacity, and minimal bleed for sharp peaks.
  • Adaptable for mass spectrometry applications; C18 ligand offers high retention for basic compounds.
  • Withstands maximum pressure of 6000 psi (415 Bar) with superior low pH stability.
  • Features proprietary silica synthesis, bonding, and end-capping procedures, improving peak shape and symmetry for resolution and quantitation of acidic, neutral, and basic compounds at 2 to 8 pH range.