Learn More
Waters Corp Waters Column, C18, 5 μm, 3 mm ID, 50 mm Length
Supplier:  Waters Corp 186002545
Encompass_Preferred
SunFire C18 columns are designed for general-purpose method development at low pH. Their C18 ligand provides high retention, especially for basic compounds, making them suitable for purification and impurity profile assays. These columns deliver quick, low-back pressure, and high-resolution performance, adaptable for various mass spectrometry applications.
  • Designed for general purpose method development at low pH.
  • C18 ligand offers high retention for basic compounds.
  • Ideal for purification and impurity profile assays.
  • Provides quick, low-back pressure, and high-resolution separation.
  • Ensures strong peaks, outstanding sensitivity, and negligible bleed.
  • Adaptable for numerous mass spectrometry applications.
  • Manufactured in a cGMP, ISO 9001-certified laboratory with ultra-pure reagents.
  • Each batch chromatographically tested for repeatable performance.
Catalog No. 50-781-185