Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp SunFire C18 Column, 100Å, 5 μm, 4.6 mm × 250 mm

Catalog No. 50781200 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This C18 column is designed for reversed-phase high-performance liquid chromatography (HPLC) applications.

  • Features a 100Å pore size and 5 μm particle size, with 4.6 mm inner diameter and 250 mm length.
  • Used for general purpose method development at low pH.
  • Provides high retention, especially for basic compounds, suitable for purification and impurity profile assays.
  • Designed to handle complex gradient chromatography applications.
  • Delivers sharp peaks and consistent performance for excellent peak shape and resolution.
  • Provides reproducibility and scalability for method development.
  • Features a pH range from 2 to 8.
  • Withstands a maximum pressure of 6000 psi (415 Bar).
  • Spherical silica particles with low silanol activity.
  • Endcapped, with 16% carbon load and 340 surface area.
  • Classified as USP L1.

Catalog No. 50-781-200 Supplier Waters Corp Supplier No. 186002560
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This C18 column is designed for reversed-phase high-performance liquid chromatography (HPLC) applications.

  • Features a 100Å pore size and 5 μm particle size, with 4.6 mm inner diameter and 250 mm length.
  • Used for general purpose method development at low pH.
  • Provides high retention, especially for basic compounds, suitable for purification and impurity profile assays.
  • Designed to handle complex gradient chromatography applications.
  • Delivers sharp peaks and consistent performance for excellent peak shape and resolution.
  • Provides reproducibility and scalability for method development.
  • Features a pH range from 2 to 8.
  • Withstands a maximum pressure of 6000 psi (415 Bar).
  • Spherical silica particles with low silanol activity.
  • Endcapped, with 16% carbon load and 340 surface area.
  • Classified as USP L1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.