Learn More
Waters Corp SunFire C18 Column, 100Å, 5 µm, 4.6 mm X 150 mm, 1/pk
Catalog No. 50781199 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds and it is ideally suited for purification and impurity profile assays.
Catalog No. 50-781-199 Supplier Waters Corp Supplier No. 186002559
SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds and it is ideally suited for purification and impurity profile assays.