Learn More
Waters Corp Waters SunFire C18 Column, 5 μm, 4.6 mm ID, 50 mm Length
Supplier:  Waters Corp 186002557
Encompass_Preferred
SunFire C18 columns are ideal for general purpose method development at low pH, offering high retention for basic compounds and optimal particle and bonding expertise for superior chromatographic performance in purification and impurity profile assays.
  • Extended column lifetime due to higher low pH stability compared to conventional HPLC columns.
  • Efficient and reliable analytical tool offering increased sensitivity.
  • Utilizes proprietary silica synthesis, bonding, end-capping, and packing technologies to enable superior peak shape performance.
  • Achieves symmetrical peaks for improved resolution and quantification of acidic, neutral, and basic chemicals across pH ranges.
  • Features low back pressure, excellent resolution, quick response, superb sensitivity, high peak capacity, and minimal bleed for crisp peaks.
  • Ideal for various mass spectrometry applications.
Catalog No. 50-781-197