Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp SunFire Preparative Column, C18, 100Å, 5 μm, 10 mm × 250 mm

Catalog No. 502038365 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This C18 OBD Preparative Column is ideal for general purpose method development at low pH, offering high retention for basic compounds and suitability for purification and impurity profile assays. It separates hydrophobic and moderately polar compounds and tolerates higher pH for flexible method development.

  • C18 chemistry for reversed phase separations.
  • Spherical silica particle with 5 μm particle size and 100 Å pore size.
  • Dimensions: 10 mm inner diameter × 250 mm length.
  • pH range: 2 to 8.
  • Temperature limit: 45 °C.
  • Maximum pressure: 6000 psi (415 Bar).
  • Features low silanol activity and is endcapped.
  • Carbon load: 16%.
  • USP classification: L1.
  • Provides balanced retention for acids, bases, and neutrals.
  • Offers excellent loading capacity.
  • Stable across wide pH and temperature ranges.

Catalog No. 50-203-8365 Supplier Waters Corp Supplier No. 186008155
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This C18 OBD Preparative Column is ideal for general purpose method development at low pH, offering high retention for basic compounds and suitability for purification and impurity profile assays. It separates hydrophobic and moderately polar compounds and tolerates higher pH for flexible method development.

  • C18 chemistry for reversed phase separations.
  • Spherical silica particle with 5 μm particle size and 100 Å pore size.
  • Dimensions: 10 mm inner diameter × 250 mm length.
  • pH range: 2 to 8.
  • Temperature limit: 45 °C.
  • Maximum pressure: 6000 psi (415 Bar).
  • Features low silanol activity and is endcapped.
  • Carbon load: 16%.
  • USP classification: L1.
  • Provides balanced retention for acids, bases, and neutrals.
  • Offers excellent loading capacity.
  • Stable across wide pH and temperature ranges.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.