Learn More
Waters Corp Waters SunFire OBD Prep Column, C18, 100Å, 5 μm, 10 mm x 100 mm
Catalog No. 502038363 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds and is ideally suited for purification and impurity profile assays.
  • Used for general purpose method development at low pH.
  • C18 ligand provides high retention, especially for basic compounds.
  • Suitable for purification and impurity profile assays.
Catalog No. 50-203-8363 Supplier Waters Corp Supplier No. 186008153
These C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds and is ideally suited for purification and impurity profile assays.
  • Used for general purpose method development at low pH.
  • C18 ligand provides high retention, especially for basic compounds.
  • Suitable for purification and impurity profile assays.