Learn More
Waters Corp SunFire C18 Sentry Guard Cartridge, 100Å, 3.5 µm, 2.1 mm X 10 mm, 2/pk
Supplier:  Waters Corp 186002530
Encompass_Preferred
SunFire C18 columns are used for general purpose method development at low pH. The C18 ligand is highly retentive, especially for basic compounds and it is ideally suited for purification and impurity profile assays.
Catalog No. 50-781-170