Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Sentry Guard Cartridge, C18, 5 μm, 3 mm × 20 mm

Catalog No. 50781287 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

SunFire C18 columns are designed for general purpose method development at low pH. They feature a highly retentive C18 ligand, making them particularly suitable for separating basic compounds. These cartridges are ideally used for purification and impurity profile assays.

  • C18 chemistry for diverse applications
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 415 bar (6000 psi)
  • Endcapped for enhanced performance
  • Low silanol activity
  • Spherical particle shape
  • Particle size of 5 μm
  • Pore size of 100 Å
  • Sentry Guard Cartridge format
  • Surface area of 340
  • HPLC system compatibility
  • USP classification L1
  • Inner diameter of 3 mm
  • Length of 20 mm
  • Carbon load of 16%
  • Package contains 2 units

Catalog No. 50-781-287 Supplier Waters Corp Supplier No. 186002683
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SunFire C18 columns are designed for general purpose method development at low pH. They feature a highly retentive C18 ligand, making them particularly suitable for separating basic compounds. These cartridges are ideally used for purification and impurity profile assays.

  • C18 chemistry for diverse applications
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 415 bar (6000 psi)
  • Endcapped for enhanced performance
  • Low silanol activity
  • Spherical particle shape
  • Particle size of 5 μm
  • Pore size of 100 Å
  • Sentry Guard Cartridge format
  • Surface area of 340
  • HPLC system compatibility
  • USP classification L1
  • Inner diameter of 3 mm
  • Length of 20 mm
  • Carbon load of 16%
  • Package contains 2 units

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.