Learn More
Waters Corp SunFire VanGuard Cartridge, C18, 100 Å, 5 μm, 3.9 mm x 5 mm
Catalog No. 502391188 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C18 VanGuard Cartridges are designed to extend analytical column lifetime and performance by removing particulate and chemical contamination. This cartridge is installed between the injector and analytical column, ensuring compatibility and optimal protection for various HPLC, UPLC, and UHPLC systems.
  • Extends analytical column lifetime and performance.
  • Removes particulates and chemical contamination.
  • Optimized for 4.6 mm I.D. analytical columns with 5 μm sorbent particles.
  • Uses matching packing material for seamless compatibility.
  • Manufactured in a cGMP, ISO 9001 certified plant with tested batches ensuring reproducible performance.
  • Improves selectivity, sensitivity, and analysis speed.
  • Direct-attach, single-unit design minimizes extra volume and reduces leaking.
  • Cost-effective protection for 2.1 mm, 3.0 mm, 3.9 mm, and 4.6 mm I.D. analytical columns.
Catalog No. 50-239-1188 Supplier Waters Corp Supplier No. 186007699
SunFire C18 VanGuard Cartridges are designed to extend analytical column lifetime and performance by removing particulate and chemical contamination. This cartridge is installed between the injector and analytical column, ensuring compatibility and optimal protection for various HPLC, UPLC, and UHPLC systems.
  • Extends analytical column lifetime and performance.
  • Removes particulates and chemical contamination.
  • Optimized for 4.6 mm I.D. analytical columns with 5 μm sorbent particles.
  • Uses matching packing material for seamless compatibility.
  • Manufactured in a cGMP, ISO 9001 certified plant with tested batches ensuring reproducible performance.
  • Improves selectivity, sensitivity, and analysis speed.
  • Direct-attach, single-unit design minimizes extra volume and reduces leaking.
  • Cost-effective protection for 2.1 mm, 3.0 mm, 3.9 mm, and 4.6 mm I.D. analytical columns.