Learn More
Waters Corp SunFire C18 VanGuard Cartridge, 100Ã…, 3.5 µm, 3.9 mm X 5 mm, 3/pk;
Catalog No. 502038159 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C18 VanGuard Cartridge, 100Å, 3.5 µm, 3.9 mm X 5 mm, 3/pk;
Catalog No. 50-203-8159 Supplier Waters Corp Supplier No. 186007696
SunFire C18 VanGuard Cartridge, 100Å, 3.5 µm, 3.9 mm X 5 mm, 3/pk;