Learn More
Waters Corp SunFire C8 Column, 100 Å, 5 μm, 4.6 mm × 150 mm
Catalog No. 502391624 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This SunFire C8 Column features a C8 stationary phase, 100 Å pore size, 5 μm particle size, and 4.6 mm × 150 mm dimensions. It includes an eConnect Column Tag that stores unique product information for seamless data integration with Empower CDS and easy web access via NFC.
  • eConnect Tag comes pre-equipped, containing identifying information to simplify setup and reduce manual data entry.
  • The permanent, tamper-proof eConnect Tag aids in compliance, system setup, and automatic column detection.
  • Fit-for-purpose tag design improves detection probability and minimizes interference in temperature-sensitive LC methods.
  • NFC capability provides convenient access to column information and knowledge on the Waters website.
  • Identical quality standards and equivalent performance to untagged counterparts are ensured.
Catalog No. 50-239-1624 Supplier Waters Corp Supplier No. 186002737RF
This SunFire C8 Column features a C8 stationary phase, 100 Å pore size, 5 μm particle size, and 4.6 mm × 150 mm dimensions. It includes an eConnect Column Tag that stores unique product information for seamless data integration with Empower CDS and easy web access via NFC.
  • eConnect Tag comes pre-equipped, containing identifying information to simplify setup and reduce manual data entry.
  • The permanent, tamper-proof eConnect Tag aids in compliance, system setup, and automatic column detection.
  • Fit-for-purpose tag design improves detection probability and minimizes interference in temperature-sensitive LC methods.
  • NFC capability provides convenient access to column information and knowledge on the Waters website.
  • Identical quality standards and equivalent performance to untagged counterparts are ensured.