Learn More
Waters Corp SunFire C8 Column, Reversed-Phase, 100 Å, 5 μm
Catalog No. 502390802 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C8 columns are designed for general purpose method development, particularly at low pH. The C8 ligand, being less hydrophobic than C18, offers reduced retention for certain assays. This column is equipped with an electronic eConnect column tag, which contains unique, column-specific information, enabling automatic identification, tracking, and digital tracing of its use history. While compatible with any HPLC system, the full benefits of eConnect columns are realized when paired with the Waters Alliance iS System running Empower software.
  • Used for general purpose method development at low pH.
  • C8 ligand is less hydrophobic and less retentive compared to C18.
  • Equipped with eConnect column tag for automatic identification, tracking, and digital tracing.
  • Compatible with any HPLC system.
  • Optimized for use with the Waters Alliance iS System running Empower software.
Catalog No. 50-239-0802 Supplier Waters Corp Supplier No. 186002738RF
SunFire C8 columns are designed for general purpose method development, particularly at low pH. The C8 ligand, being less hydrophobic than C18, offers reduced retention for certain assays. This column is equipped with an electronic eConnect column tag, which contains unique, column-specific information, enabling automatic identification, tracking, and digital tracing of its use history. While compatible with any HPLC system, the full benefits of eConnect columns are realized when paired with the Waters Alliance iS System running Empower software.
  • Used for general purpose method development at low pH.
  • C8 ligand is less hydrophobic and less retentive compared to C18.
  • Equipped with eConnect column tag for automatic identification, tracking, and digital tracing.
  • Compatible with any HPLC system.
  • Optimized for use with the Waters Alliance iS System running Empower software.