Learn More
Waters Corp Waters SunFire C8 Column, 100Å, 2.5 μm, 2.1 mm x 75 mm
Supplier: Waters Corp 186005635
SunFire C8 columns are designed for general purpose method development at low pH, suitable for specific assays and mass spectrometry applications. They offer sharp peaks, high sensitivity, and excellent resolution, providing efficient and reliable chromatographic results with outstanding repeatability. Their low pH stability extends column lifespan, and high efficiency is achieved through patented silica synthesis processes.
- C8 chemistry
- Reversed phase separation
- Silica particle
- 2.5 μm particle size
- 100 Å pore size
- 2.1 mm inner diameter
- 75 mm length
- pH range: 2 to 8
- Max pressure: 415 bar (6,000 psi)
- HPLC system
- Sharp peaks
- High sensitivity
- High peak capacity
- Low bleed
- Excellent resolution
- Reduced backpressure
- Outstanding repeatability
- Low pH stability for longer lifespan
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.