Learn More
Waters Corp Waters Column, C8, 3.5 μm Particle, 3 mm x 150 mm
Catalog No. 50781312 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to C18 columns.
  • Used for general purpose method development at low pH
  • C8 ligand is less hydrophobic and less retentive compared to C18 for certain assays
Catalog No. 50-781-312 Supplier Waters Corp Supplier No. 186002721
Add to Cart
Add to Cart
These C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to C18 columns.
  • Used for general purpose method development at low pH
  • C8 ligand is less hydrophobic and less retentive compared to C18 for certain assays