Learn More
Waters Corp Waters Column, C8, 3.5 μm, 100 Å, 3 mm x 50 mm
Catalog No. 50781310 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C8 columns are utilized for general purpose method development at low pH. The C8 ligand is favored for certain assays due to its reduced hydrophobicity, making it less retentive compared to SunFire C18.
  • C8 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range from 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 3.5 μm particle size
  • 100 Å pore size
  • HPLC system compatible
  • L7 USP classification
  • 3 mm inner diameter
  • 50 mm length
  • 12% carbon load
  • 1 unit per package
Catalog No. 50-781-310 Supplier Waters Corp Supplier No. 186002719
SunFire C8 columns are utilized for general purpose method development at low pH. The C8 ligand is favored for certain assays due to its reduced hydrophobicity, making it less retentive compared to SunFire C18.
  • C8 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range from 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 3.5 μm particle size
  • 100 Å pore size
  • HPLC system compatible
  • L7 USP classification
  • 3 mm inner diameter
  • 50 mm length
  • 12% carbon load
  • 1 unit per package