Learn More
Waters Corp Waters SunFire C8 Column, 100 Å, 3.5 μm, 4.6x100 mm
Supplier: Waters Corp 186002731
SunFire C8 columns are designed for general purpose method development at low pH, offering less hydrophobicity and retention than C18 columns. They are suitable for various mass spectrometry applications, delivering consistent chromatographic results and exceptional repeatability for pharmaceutical and biopharmaceutical analyses. These columns combine patented silica synthesis, bonding, and packing processes for high efficiency, increased sensitivity, and longer lifetime due to low pH stability. They can withstand up to 6000 psi (415 Bar) pressure.
- Sharp peaks
- Remarkable sensitivity
- High peak capacity
- Ultra-low bleed
- Great resolution
- Minimal backpressure
- Speed
- Longer column lifetime due to low pH stability