Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters SunFire C8 Column, 100Å, 3.5 μm, 4.6 mm X 75 mm

Catalog No. 50781321 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

SunFire C8 columns are designed for general purpose method development in low pH environments. The C8 ligand is preferred for certain assays due to its reduced hydrophobicity, resulting in less retention compared to SunFire C18 columns.

  • Suitable for general purpose method development at low pH.
  • C8 ligand offers less hydrophobicity and retention compared to SunFire C18.
  • Chemistry: C8
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • Particle size: 3.5 μm
  • Pore size: 100 Å
  • Inner diameter: 4.6 mm
  • Length: 75 mm
  • System: HPLC
  • pH range: 2 to 8
  • Maximum pressure: 6000 psi (415 Bar)

Catalog No. 50-781-321 Supplier Waters Corp Supplier No. 186002730
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SunFire C8 columns are designed for general purpose method development in low pH environments. The C8 ligand is preferred for certain assays due to its reduced hydrophobicity, resulting in less retention compared to SunFire C18 columns.

  • Suitable for general purpose method development at low pH.
  • C8 ligand offers less hydrophobicity and retention compared to SunFire C18.
  • Chemistry: C8
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • Particle size: 3.5 μm
  • Pore size: 100 Å
  • Inner diameter: 4.6 mm
  • Length: 75 mm
  • System: HPLC
  • pH range: 2 to 8
  • Maximum pressure: 6000 psi (415 Bar)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.