Learn More
Waters Corp Waters Column, C8, 3.5 μm, 4.6 mm X 50 mm, 100 Å
Catalog No. 50781320 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed for general purpose method development at low pH. It offers high efficiency and enhanced sensitivity, utilizing a C8 ligand that is less hydrophobic and less retentive than C18, making it suitable for specific assays. Its excellent low pH stability ensures a longer column lifetime compared to other silica-based HPLC columns.
  • Ideal for low pH method development.
  • C8 ligand for less hydrophobic, less retentive separations.
  • Extended column lifetime from low pH stability.
  • High efficiency and sensitivity via patented silica synthesis.
  • Supports mass spectrometry; delivers sharp peaks, low bleed.
  • Achieves excellent resolution, reduced back pressure, and speed.
  • Reliable and reproducible chromatographic results.
  • Manufactured with high-purity materials under strict quality control.
Catalog No. 50-781-320 Supplier Waters Corp Supplier No. 186002729
Add to Cart
Add to Cart
This column is designed for general purpose method development at low pH. It offers high efficiency and enhanced sensitivity, utilizing a C8 ligand that is less hydrophobic and less retentive than C18, making it suitable for specific assays. Its excellent low pH stability ensures a longer column lifetime compared to other silica-based HPLC columns.
  • Ideal for low pH method development.
  • C8 ligand for less hydrophobic, less retentive separations.
  • Extended column lifetime from low pH stability.
  • High efficiency and sensitivity via patented silica synthesis.
  • Supports mass spectrometry; delivers sharp peaks, low bleed.
  • Achieves excellent resolution, reduced back pressure, and speed.
  • Reliable and reproducible chromatographic results.
  • Manufactured with high-purity materials under strict quality control.