Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters SunFire C8 Column, 5 μm, 2.1 mm, 150 mm

Catalog No. 50781308 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

SunFire C8 columns are utilized for general-purpose method development under low pH conditions. The C8 ligand is favored for certain assays due to its lower hydrophobicity, which results in less retention compared to SunFire C18 columns.

  • C8 chemistry for general purpose method development.
  • Separation mode: Reversed phase.
  • Particle substrate: Silica.
  • pH range: 2 pH to 8 pH.
  • Maximum pressure: 6000 psi (415 Bar).
  • Endcapped.
  • Low silanol activity.
  • Particle shape: Spherical.
  • Particle size: 5 μm.
  • Pore size: 100 Å.
  • Designed for HPLC systems.
  • Inner diameter: 2.1 mm.
  • Length: 150 mm.
  • Carbon load: 12 %.
  • Units per package: 1.

Catalog No. 50-781-308 Supplier Waters Corp Supplier No. 186002717
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SunFire C8 columns are utilized for general-purpose method development under low pH conditions. The C8 ligand is favored for certain assays due to its lower hydrophobicity, which results in less retention compared to SunFire C18 columns.

  • C8 chemistry for general purpose method development.
  • Separation mode: Reversed phase.
  • Particle substrate: Silica.
  • pH range: 2 pH to 8 pH.
  • Maximum pressure: 6000 psi (415 Bar).
  • Endcapped.
  • Low silanol activity.
  • Particle shape: Spherical.
  • Particle size: 5 μm.
  • Pore size: 100 Å.
  • Designed for HPLC systems.
  • Inner diameter: 2.1 mm.
  • Length: 150 mm.
  • Carbon load: 12 %.
  • Units per package: 1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.