Learn More
Waters Corp SunFire C8 Column, 100Å, 5 µm, 2.1 mm X 100 mm, 1/pk
Catalog No. 50781307 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.
Catalog No. 50-781-307 Supplier Waters Corp Supplier No. 186002716
SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.