Learn More
Waters Corp SunFire C8 Column, 100Å, 5 μm, 3 mm x 150 mm
Supplier:  Waters Corp 186002725
Encompass_Preferred
Waters SunFire HPLC Columns are engineered for high mass loading, excellent low pH stability, superior peak shapes, and high efficiency. SunFire C8 columns are general-purpose silica columns, offering compatibility with Mass Spectrometry (MS).
  • General purpose silica column
  • High efficiency
  • MS compatible
  • Excellent low pH stability
  • Superior peak shapes
  • Exceptional loading capacity
  • Low mass spectral (MS) bleed
Catalog No. 50-781-316