Learn More
Waters Corp Waters SunFire C8 Column, 5 μm, 4.6 mm X 250 mm, 100 Å
Supplier:  Waters Corp 186002738
Encompass_Preferred
SunFire C8 columns are optimized for general purpose method development at low pH. They feature a C8 ligand that is less hydrophobic and retentive than C18, making them suitable for specific assays. Advanced bonding and end-capping technologies provide superior peak shape, enhancing resolution and quantitation of various compounds across low to intermediate pH ranges.
  • Delivers symmetrical peaks.
  • Stable at low pH, extending column lifetime.
  • Suitable for mass spectrometry applications.
  • Provides sharp peaks, excellent sensitivity, and ultra-low bleed.
  • Achieves high efficiency with proprietary technologies.
  • Manufactured in an ISO 9000 certified facility.
  • Each batch undergoes chromatographic testing for reproducible performance.
  • Supplied with a Performance Chromatogram and Certificate of Batch Analysis.
Catalog No. 50-781-329