Learn More
Waters Corp Waters HPLC Column, C8, 5 μm, 4.6 mm x 50 mm
Supplier:  Waters Corp 186002735
Encompass_Preferred
These C8 columns are designed for general purpose method development at low pH, offering high efficiency and increased sensitivity. Less hydrophobic than C18 columns, they provide a longer column lifetime due to low pH stability. They are suitable for mass spectrometry applications, ensuring consistent chromatographic results and reproducibility.
  • General purpose method development at low pH.
  • Less hydrophobic and retentive than C18 columns, with longer lifetime due to low pH stability.
  • High efficiency and increased sensitivity for mass spectrometry applications.
  • Delivers sharp peaks, high sensitivity, high peak capacity, and very low bleed.
  • Excellent resolution, low back pressure, and speed.
  • Consistent chromatographic results and reproducibility.
  • Manufactured with high-purity raw materials and strict quality control.
Catalog No. 50-781-326