Learn More
Waters Corp SunFire Column, Method Validation Kit, C8 Silica, 3.5 μm
Supplier:  Waters Corp 186002740
Encompass_Preferred
This kit consists of three C8 columns packed with different batches of sorbent, offering low pH stability for extended column lifetime. It integrates patented silica synthesis, bonding, end-capping, and packing techniques for high efficiency and improved sensitivity.
  • Suitable for a wide range of mass spectrometry applications.
  • Provides sharp peaks, exceptional sensitivity, large peak capacity, and ultra-low bleed.
  • Offers high resolution, low back pressure, and fast analysis.
  • Delivers consistent chromatographic results and excellent repeatability year after year, batch after batch, and column after column.
  • Manufactured using high-purity raw materials under tight quality control and packing procedures.
Catalog No. 50-781-331