Learn More
Waters Corp Waters Corporation Sentry Guard Cartridge, C8, 3.5 μm, 100 Å, 4.6 mm X 20 mm
Catalog No. 50781318 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.
  • C8 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 3.5 μm particle size
  • 100 Å pore size
  • Sentry guard cartridge format
  • 340 surface area
  • HPLC system compatibility
  • L7 USP classification
  • 4.6 mm inner diameter
  • 20 mm length
  • 12% carbon load
  • 2 units per package
Catalog No. 50-781-318 Supplier Waters Corp Supplier No. 186002727
SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.
  • C8 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 3.5 μm particle size
  • 100 Å pore size
  • Sentry guard cartridge format
  • 340 surface area
  • HPLC system compatibility
  • L7 USP classification
  • 4.6 mm inner diameter
  • 20 mm length
  • 12% carbon load
  • 2 units per package