Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp SunFire Sentry Guard Cartridge, C8, 3.5 μm, 3 mm × 20 mm

Catalog No. 50781309 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

SunFire C8 Sentry Guard Cartridges are designed for general-purpose method development in low pH conditions. Their C8 ligand provides lower hydrophobicity and reduced retention compared to C18 phases, making them suitable for specific assays.

  • Designed for general-purpose method development in low pH conditions.
  • Features C8 ligand for lower hydrophobicity and reduced retention.
  • Separation mode: Reversed Phase.
  • Particle substrate: Silica.
  • pH range: 2 to 8.
  • Maximum pressure: 6000 psi (415 Bar).
  • Endcapped for enhanced performance.
  • Low silanol activity.
  • Spherical particle shape.
  • Particle size: 3.5 μm.
  • Pore size: 100 Å.
  • Format: Sentry Guard Cartridge.
  • System compatibility: HPLC.
  • USP classification: L7.
  • Inner diameter: 3 mm.
  • Length: 20 mm.
  • Carbon load: 12%.
  • Units per package: 2.

Catalog No. 50-781-309 Supplier Waters Corp Supplier No. 186002718
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SunFire C8 Sentry Guard Cartridges are designed for general-purpose method development in low pH conditions. Their C8 ligand provides lower hydrophobicity and reduced retention compared to C18 phases, making them suitable for specific assays.

  • Designed for general-purpose method development in low pH conditions.
  • Features C8 ligand for lower hydrophobicity and reduced retention.
  • Separation mode: Reversed Phase.
  • Particle substrate: Silica.
  • pH range: 2 to 8.
  • Maximum pressure: 6000 psi (415 Bar).
  • Endcapped for enhanced performance.
  • Low silanol activity.
  • Spherical particle shape.
  • Particle size: 3.5 μm.
  • Pore size: 100 Å.
  • Format: Sentry Guard Cartridge.
  • System compatibility: HPLC.
  • USP classification: L7.
  • Inner diameter: 3 mm.
  • Length: 20 mm.
  • Carbon load: 12%.
  • Units per package: 2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.