Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Sentry Guard Cartridge, C8, 100Å, 5 μm, 2.1 mm X 10 mm

Catalog No. 50781304 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.

  • C8 chemistry for general purpose method development at low pH.
  • Less hydrophobic and less retentive C8 ligand compared to C18, suitable for specific assays.
  • Spherical silica particle substrate.
  • Particle size of 5 μm and pore size of 100 Å.
  • Designed for HPLC systems.
  • Maximum pressure rating of 6000 psi (415 bar).
  • Available in a 2-pack.

Catalog No. 50-781-304 Supplier Waters Corp Supplier No. 186002713
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SunFire C8 columns are used for general purpose method development at low pH. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to SunFire C18.

  • C8 chemistry for general purpose method development at low pH.
  • Less hydrophobic and less retentive C8 ligand compared to C18, suitable for specific assays.
  • Spherical silica particle substrate.
  • Particle size of 5 μm and pore size of 100 Å.
  • Designed for HPLC systems.
  • Maximum pressure rating of 6000 psi (415 bar).
  • Available in a 2-pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.