Learn More
Waters Corp VanGuard Cartridge SunFire C8, 3.5 μm, 2.1 x 5 mm
Catalog No. 502391841 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The SunFire C8 VanGuard Cartridge is designed to protect analytical columns by capturing particulate matter and chemical contaminants. It features a SunFire C8 phase with a 3.5 μm particle size and 100 Å pore size, ensuring efficient sample preparation and extended column lifetime. This guard cartridge measures 2.1 mm ID by 5 mm in length.
  • Guard column type
  • SunFire C8 phase
  • 3.5 μm particle size
  • 100 Å pore size
  • 2.1 mm ID
  • 5 mm length
  • USP L7 compliant
Catalog No. 50-239-1841 Supplier Waters Corp Supplier No. 186007703
The SunFire C8 VanGuard Cartridge is designed to protect analytical columns by capturing particulate matter and chemical contaminants. It features a SunFire C8 phase with a 3.5 μm particle size and 100 Å pore size, ensuring efficient sample preparation and extended column lifetime. This guard cartridge measures 2.1 mm ID by 5 mm in length.
  • Guard column type
  • SunFire C8 phase
  • 3.5 μm particle size
  • 100 Å pore size
  • 2.1 mm ID
  • 5 mm length
  • USP L7 compliant