Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation SunFire Prep Guard Cartridge, C18, 5 μm, 10 x 10 mm

Catalog No. 50781205 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These C18 prep guard cartridges are designed for general purpose method development at low pH. The C18 ligand provides high retention, particularly for basic compounds, making them suitable for purification and impurity profile assays.

  • C18 chemistry for high retention
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Temperature limit of 45°C
  • Maximum pressure of 6000 psi (415 bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • Particle size of 5 μm
  • Pore size of 100 Å
  • HPLC system compatibility
  • Suitable for LC/MS and LC techniques
  • USP classification L1
  • Inner diameter of 10 mm
  • Length of 10 mm
  • Carbon load of 16%
  • One unit per package

Catalog No. 50-781-205 Supplier Waters Corp Supplier No. 186002565
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These C18 prep guard cartridges are designed for general purpose method development at low pH. The C18 ligand provides high retention, particularly for basic compounds, making them suitable for purification and impurity profile assays.

  • C18 chemistry for high retention
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Temperature limit of 45°C
  • Maximum pressure of 6000 psi (415 bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • Particle size of 5 μm
  • Pore size of 100 Å
  • HPLC system compatibility
  • Suitable for LC/MS and LC techniques
  • USP classification L1
  • Inner diameter of 10 mm
  • Length of 10 mm
  • Carbon load of 16%
  • One unit per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.