Learn More
Waters Corp Waters SunFire C18 Prep Guard Cartridge, 5 μm, 100 Å, 19 mm × 10 mm
Catalog No. 50781209 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This SunFire C18 Prep Guard Cartridge is a single piece of lab equipment for general purpose method development at low pH. Its C18 ligand is highly retentive for basic compounds. It protects columns by removing particle pollution, extending their lifetime and ensuring compatibility with UPLC columns for reversed-phase Liquid Chromatography.
  • Extends prep column life, preserves analytical performance.
  • Longer column lifetime, lower purification expenses.
  • Protects C18 columns, extends lab equipment life by removing particle pollution.
  • Protects 19 mm I.D. C18 columns with 5 μm sorbent particles.
  • Tolerates high pressures in UPLC system applications.
  • Delivers optimal performance across a wide operating pH range.
  • Includes patented BEH Technology C18, 100Å, 5μm particles.
  • Q.C. tested to ensure consistent performance, longer analytical column life.
Catalog No. 50-781-209 Supplier Waters Corp Supplier No. 186002569
This SunFire C18 Prep Guard Cartridge is a single piece of lab equipment for general purpose method development at low pH. Its C18 ligand is highly retentive for basic compounds. It protects columns by removing particle pollution, extending their lifetime and ensuring compatibility with UPLC columns for reversed-phase Liquid Chromatography.
  • Extends prep column life, preserves analytical performance.
  • Longer column lifetime, lower purification expenses.
  • Protects C18 columns, extends lab equipment life by removing particle pollution.
  • Protects 19 mm I.D. C18 columns with 5 μm sorbent particles.
  • Tolerates high pressures in UPLC system applications.
  • Delivers optimal performance across a wide operating pH range.
  • Includes patented BEH Technology C18, 100Å, 5μm particles.
  • Q.C. tested to ensure consistent performance, longer analytical column life.